Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Green Fluorescent Protein Rabbit anti-Vertebrates, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB3080PMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB3080PMI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB3080PMI Supplier MilliporeSigma Supplier No. AB3080P
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Green Fluorescent Protein Antibody detects level of Green Fluorescent Protein and has been published and validated for use in ELISA, IC and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Green Fluorescent Protein
Applications ELISA, Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified rabbit polyclonal IgG in 10mM Tris buffer.
Gene Symbols GFP; eGFP
Host Species Rabbit
Immunogen Highly purified native GFP from Aequorea victoria.
Purification Method Affinity Purified
Quantity 50 μg
Primary or Secondary Primary
Target Species Rat, Vertebrates
Content And Storage -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.