Learn More
Description
Specifications
Specifications
| Antigen | GFP |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 3F8.2 |
| Conjugate | Unconjugated |
| Formulation | Purified mouse immunoglobulin in buffer containing PBS containing 0.05% sodium azide. |
| Gene Symbols | GFP |
| Host Species | Mouse |
| Immunogen | Full length GFP protein. |
| Purification Method | Protein G purfied |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.