Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GGA3 Mouse anti-Human, Unlabeled, Clone: 8, BD

Catalog No. BDB612311 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612311 150 μg
BDB612310 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB612311 Supplier BD Biosciences Supplier No. 612311
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The ADP-ribosylation factors (ARFs) are a family of small GTPases in the ARF superfamily that include ARFs and ARF-like (ARLs) proteins. At least six ARFs have been identified in humans: ARF1, ARF2, ARF3, ARF4, ARF5, and ARF6. ARFs are involved in intravesicular acidification and fusion of microsomal vesicles, endosome fusion, nuclear membrane assembly, and formation of clathrin-coated vesicles. GGAs are ARF-binding proteins that act as adaptor coat proteins associated with the Golgi complex. GGA1, GGA2, and GGA3 are homologous proteins that contain N-terminal VHS domains, a GGA and TOM homology region (GAT), and a C-terminal region homologous to the ear domain of γ-adaptins. GGAs co-localize with Golgi markers in the TGN, and GGA3 is found present in coated vesicles and buds associated with the TGN. The GAT domain of GGA3 facilitates ARF1 binding, Golgi localization, and dissociation from ARF-regulated membranes. The C-terminal region of GGAs bind to MAP1A and rabaptin-5, which are binding partners of γ-adaptins. Overexpression of GGAs alters the distribution of markers normally found in the TGN. Thus, GGAs are ARF binding proteins that regulate vesicle dynamics in the TGN.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen GGA3
Applications Western Blot
Classification Monoclonal
Clone 8
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human GGA3 aa. 424-542
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.