Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GGCT Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA526679
Description
GGCT Polyclonal Antibody for Western Blot
GGCT (EC 2.3.2.4) catalyzes the formation of 5-oxoproline (pyroglutamic acid) from gamma-glutamyl dipeptides and may play a significant role in glutathione homeostasis.
Specifications
| GGCT | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| O75223 | |
| GGCT | |
| KLH conjugated synthetic peptide between 139-168 amino acids from the C-terminal region of human GGCT. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| GGCT | |
| A030007L17Rik; AA673177; C7orf24; CRF21; Cytochrome c-releasing factor 21; gamma -glutamyl cyclotransferase; gamma-glutamyl cyclotransferase; gamma-glutamyl cyclotransferase-like 1; gamma-glutamylcyclotransferase; Gctg; GGC; GGCT; Ggctl1; RGD1304876; RGD1562590 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 79017 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction