Learn More
GGT1, Mouse, Clone: 1F9, Abnova™
Mouse monoclonal antibody raised against a partial recombinant GGT1.
Supplier: Abnova Corporation H00002678M01
Description
The enzyme encoded by this gene catalyzes the transfer of the glutamyl moiety of glutathione to a variety of amino acids and dipeptide acceptors. The enzyme is composed of a heavy chain and a light chain, which are derived from a single precursor protein, and is present in tissues involved in absorption and secretion. This enzyme is a member of the gamma-glutamyltransferase protein family, of which many members have not yet been fully characterized and some of which may represent pseudogenes. This gene is classified as type I gamma-glutamyltransferase. Multiple alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq
Sequence: TAHLSVVAEDGSAVSATSTINLYFGSKVRSPVSGILFNNEMDDFSSPSITNEFGVPPSPANFIQPGKQPLSSMCPTIMVGQDGQVRMVVGSpecifications
GGT1 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_005265 | |
GGT1 | |
GGT1 (NP_005256, 381 a.a. ∼ 470 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2a κ |
ELISA, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot | |
1F9 | |
Mouse monoclonal antibody raised against a partial recombinant GGT1. | |
GGT1 | |
CD224/D22S672/D22S732/GGT/GTG/MGC96892/MGC96904/MGC96963 | |
Mouse | |
Affinity chromatography | |
RUO | |
2678 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.