Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GH2, Mouse, Clone: 1E11, Abnova™

Catalog No. 89022307 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-022-307 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-022-307 Supplier Abnova Corporation Supplier No. H00002689M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant GH2.

The protein encoded by this gene is a member of the somatotropin/prolactin family of hormones which play an important role in growth control. The gene, along with four other related genes, is located at the growth hormone locus on chromosome 17 where they are interspersed in the same transcriptional orientation; an arrangement which is thought to have evolved by a series of gene duplications. The five genes share a remarkably high degree of sequence identity. Alternative splicing generates additional isoforms of each of the five growth hormones, leading to further diversity and potential for specialization. As in the case of its pituitary counterpart, growth hormone 1, the predominant isoform of this particular family member shows similar somatogenic activity, with reduced lactogenic activity. Mutations in this gene lead to placental growth hormone/lactogen deficiency. [provided by RefSeq

Sequence: FPTIPLSRLFDNAMLRARRLYQLAYDTYQEFEEAYILKEQKYSFLQNPQTSLCFSESIPTPSNRVKTQQKSNLELLRISLLLIQSWLEPVQLLRSVFANSLVYGASDSNVYRHLKDLEEGIQTLMWRLEDGSPRTGQIFNQSYSKFDTKSHNDDALLKNYGLLYCFRKDMDKVETFLRIVQCRSVEGSCGF

Specifications

Antigen GH2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1E11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant GH2.
Formulation PBS with no preservative; pH 7.4
Gene GH2
Gene Accession No. BC020760
Gene Alias GH-V/GHL/GHV/hGH-V
Gene Symbols GH2
Host Species Mouse
Immunogen GH2 (AAH20760.1, 27 a.a. ∼ 217 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2689
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.