Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ghrelin Mouse anti-Mouse, Human, Clone: 1ML-1D7, MilliporeSigma™

Catalog No. MAB10404MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB10404MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB10404MI Supplier MilliporeSigma Supplier No. MAB10404
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Ghrelin using this Anti-Ghrelin Antibody, clone 1ML-1D7 validated for use in western blotting and immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ghrelin
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 1ML-1D7
Conjugate Unconjugated
Formulation Unpurified mouse monoclonal ascites with 0.05% sodium azide.
Gene Accession No. NP_057446.1
Gene Symbols MTLRP1, GHRL
Host Species Mouse
Immunogen Linear Peptide corresponding to the N-terminus of human Ghrelin.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.