Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GILT Mouse anti-Mouse, PE, Clone: MaP.mGILT6, BD

Catalog No. BDB564875 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564875 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564875 Supplier BD Biosciences Supplier No. 564875

Mouse Monoclonal Antibody

The MaP.mGILT6 monoclonal antibody specifically binds to mouse Gamma-interferon-inducible lysosomal thiol reductase (GILT). GILT is encoded by Interferon gamma inducible protein 30 ( Ifi30 ). This enzyme, also known as Lysosomal thiol reductase IP30, functions in the endocytic pathway by reducing protein disulfide bonds and facilitating protein unfolding. GILT is constitutively expressed at various levels in macrophages, dendritic cells, B cells, thymocytes, and T cells, and is induced by interferon gamma in other cell types. Enzymatically active GILT dimers can also be secreted by macrophages in response to bacterial stimulation. This enzyme plays an important role in MHC class II-restricted antigen processing and can promote MHC class I-restricted presentation as well.

TRUSTED_SUSTAINABILITY

Specifications

Antigen GILT
Applications Flow Cytometry
Classification Monoclonal
Clone MaP.mGILT6
Concentration 0.2mg/mL
Conjugate PE
Description Ifi30, GILT, Ip30
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse GILT Recombinant Protein
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.