Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GTPase, IMAP family member 4, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017116 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-116 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-116 Supplier Abnova Corporation Supplier No. H00055303B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human GIMAP4 protein.

This gene encodes a protein belonging to the GTP-binding superfamily and to the immuno-associated nucleotide (IAN) subfamily of nucleotide-binding proteins. The encoded protein of this gene may be negatively regulated by T-cell acute lymphocytic leukemia 1 (TAL1). In humans, the IAN subfamily genes are located in a cluster at 7q36.1. [provided by RefSeq

Sequence: MAAQYGSMSFNPSTPGASYGPGRQEPRNSQLRIVLVGKTGAGKSATGNSILGRKVFHSGTAAKSITKKCEKRSSSWKETELVVVDTPGIFDTEVPNAETSKEIIRCILLTSPGPHALLLVVPLGRYTEEEHKATEKILKMFGERARSFMILIFTRKDDLGDTNLHDYLREAPEDIQDLMDIFGDRYCALNNKATGAEQEAQRAQLLGLIQRVVRENKEGCYTNRMYQRAEEEIQKQTQAMQELHRVELEREKARIREEYEEKIRKLEDKVEQEKRKKQMEKKLAEQEAHYAVRQQRARTEVESKDGILELIMTALQIASFILLRLFAED

Specifications

Antigen GTPase, IMAP family member 4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human GIMAP4 protein.
Formulation PBS with no preservative; pH 7.4
Gene GIMAP4
Gene Accession No. NM_018326.2
Gene Alias FLJ11110/HIMAP4/IAN1/IMAP4/MSTP062/hIAN1
Gene Symbols GIMAP4
Host Species Mouse
Immunogen GIMAP4 (NP_060796.1, 1 a.a. ∼ 329 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55303
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.