Learn More
GINS2, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human GINS2 peptide using ARM Technology.
Supplier: Abnova Corporation H00051659K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
The yeast heterotetrameric GINS complex is made up of Sld5 (GINS4; MIM 610611), Psf1 (GINS1; MIM 610608), Psf2, and Psf3 (GINS3; MIM 610610). The formation of this complex is essential for the initiation of DNA replication in yeast and Xenopus egg extracts (Ueno et al., 2005 [PubMed 16287864]). See GINS1 for additional information about the GINS complex.[supplied by OMIM
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
GINS2 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
GINS2 | |
GINS2 | |
A synthetic peptide of human GINS2 | |
RUO | |
51659 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
GINS complex subunit 2 (Psf2 homolog) | |
GINS2 | |
HSPC037, PSF2, Pfs2 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.