Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Girdin, coiled-coil region Rabbit anti-Human, Monkey, Polyclonal, MilliporeSigma™

Catalog No. ABT80MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABT80MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABT80MI Supplier MilliporeSigma Supplier No. ABT80
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-Girdin, coiled-coil region Antibody is validated for use in immunocytochemistry, Western Blotting, Immunoprecipitation for the detection of Girdin, coiled-coil region.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Girdin, coiled-coil region
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Host Species Rabbit
Immunogen GST-tagged recombinant protein corresponding to the coiled coil domain of rat Girdin.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) XP_223709
Target Species Human, Monkey
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.