Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Girdin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP171857
Description
Girdin Polyclonal specifically detects Girdin in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Girdin | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
Akt phosphorylation enhancer, APEGIRDIN, coiled-coil domain containing 88A, Coiled-coil domain-containing protein 88A, G alpha-interacting vesicle-associated protein, G(alpha)-interacting vesicle-associated protein, Girders of actin filament, girdin, GIVFLJ10392, GRDNAKT-phosphorylation enhancer, HKRP1, HkRP1Galpha-interacting vesicle-associated protein, Hook-related protein 1, KIAA1212DKFZp686D0630 | |
Rabbit | |
Affinity Purified | |
RUO | |
55704 | |
Store at 4C. Do not freeze. |
Western Blot, Immunoprecipitation | |
1.0 mg/mL | |
Western Blot 1:2000-1:10000, Immunoprecipitation 2-10 ug/mg lysate | |
NP_001129069.1 | |
CCDC88A | |
The immunogen for this product maps to a region between residue 1425 and 1475 of human Coiled-Coil Domain Containing 88A using the numbering given in entry NP_001129069.1 (GeneID 55704). | |
100 μL | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction