Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GITR Rat anti-Mouse, PE, Clone: DTA-1, BD

Catalog No. BDB558119 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558119 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558119 Supplier BD Biosciences Supplier No. 558119

Rat Monoclonal Antibody

The DTA-1 antibody reacts with GITR [Glucocorticoid-induced Tumor necrosis factor (TNF) receptor family-Related], a 66-70kDa homodimer glycoprotein that is a member of the TNF receptor superfamily and is also known as TNFRSF18 (Shimuzu 2002). As its name implies, GITR expression was first detected in T lymphocytes that had been treated with dexamethasone, a glucocorticoid (Nocentini). In normal naive mice, GITR is expressed at moderate levels on CD25-positive/CD4-positive/CD8a-negative thymocytes and on CD25-positive/CD4-positive/CD45RB-low splenocytes (Shimuzu 2002, Uraushihara). It is also expressed at low levels on splenic CD25-negative/CD4-positive/CD45RB-low T lymphocytes, B lymphocytes, macrophages, and dendritic cells (Shimuzu 2002,Uraushihara). Activation of T and B lymphocytes upregulates GITR expression (Shimuzu 2002). GITR is a costimulatory receptor that plays an important role in Regulatory T (Treg)-cell functions, and a GITR Ligand has been detected on B lymphocytes, macrophages, and dendritic cells (Ji, Tone). mAb DTA-1 abrogates suppression by Treg cells without affecting their proliferative response, while it is co-stimulatory for T lymphocytes that are not Treg cells. (Bielharz, Dittmer, Kohm, Shimizu 2002, Shimizu 2003, Uraushihara).

Host Species: Rat
Clone: DTA-1
Isotype: IgG2b, λ
Cross Reactivity: Mouse

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen GITR
Applications Flow Cytometry
Classification Monoclonal
Clone DTA-1
Concentration 0.2mg/mL
Conjugate PE
Description GITR; Gitr; glucocorticoid-induced TNFR-related protein; Tnfrsf18; AITR
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse CD25+ CD4+ T Cell Line
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.