Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Connexin 46 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA511634
Description
Connexin 46 Polyclonal Antibody for Western Blot, IHC (P)
Connexin 46 (Cx46), also known as Gap Junction Protein Alpha-3 (GJA3), CAE3, CX46 and C2P3, maps to human chromosome 13q11-q12 and encodes a 46 kDa protein. Cx46, along with Cx50, is principally expressed in the lens of the eye. Cx46 mediates intercellular interactions during development and is necessary for the survival of neural crest cells. Cx46 forms gap junctions that connect lens fiber cells and are crucial for maintaining lens transparency and normal lens function. Mutations of Cx46 result in severe cataracts of the lens. Individual knockouts of Cx46 and Cx50 lead to changes in the rate of lens fiber cell differentiation and cell size, thus the interaction of Cx46 and Cx50 is required for proper organization of fiber cells.
Specifications
Connexin 46 | |
Polyclonal | |
Unconjugated | |
GJA3 | |
alpha 3 connexin; Cnx46; connexin 46; connexin-46; CTRCT14; Cx43; CX46; Cxn-46; CZP3; gap junction alpha 3; gap junction alpha-3 protein; gap junction membrane channel protein alpha 3; gap junction protein alpha 3; gap junction protein, alpha 3; gap junction protein, alpha 3, 46kDa; Gja3; Gja-3 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
2700 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q9Y6H8 | |
GJA3 | |
KLH conjugated synthetic peptide between 108-139 amino acids from the N-terminal region of human GJA3. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction