Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GJB3, Mouse, Clone: 3B4-1B3, Abnova™

Catalog No. 89022313 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-022-313 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-022-313 Supplier Abnova Corporation Supplier No. H00002707M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant GJB3.

This gene is a member of the connexin gene family. The encoded protein is a component of gap junctions, which are composed of arrays of intercellular channels that provide a route for the diffusion of low molecular weight materials from cell to cell. Mutations in this gene can cause nonsyndromic deafness or erythrokeratodermia variabilis, a skin disorder. Alternative splicing results in multiple transcript variants encoding the same protein. [provided by RefSeq

Sequence: MDWKTLQALLSGVNKYSTAFGRIWLSVVFVFRVLVYVVAAERVWGDEQKDFDCNTKQPGCTNVCYDNYFPISNIRLWALQLIFVTCPSLLVILHVAYREERERRHRQKHGDQCAKLYDNAGKKHGGLWWTYLFSLIFKLIIEFLFLYLLHTLWHGFNMPRLVQCANVAPCPNIVDCYIARPTEKKIFTYFMVGASAVCIVLTICELCYLICHRVLRGLHKDKPRGGCSPSSSASRASTCRCHHKLVEAGEVDPDPGNNKLQASAPNLTPI

Specifications

Antigen GJB3
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 3B4-1B3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant GJB3.
Formulation PBS with no preservative; pH 7.4
Gene GJB3
Gene Accession No. BC012918
Gene Alias CX31/DFNA2/EKV/FLJ22486/MGC102938
Gene Symbols GJB3
Host Species Mouse
Immunogen GJB3 (AAH12918.1, 1 a.a. ∼ 270 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2707
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.