Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glial Derived Neurotrophic Factor Chicken anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB5755MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5755MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5755MI Supplier MilliporeSigma Supplier No. AB5755
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

This Anti-Glial Derived Neurotrophic Factor Antibody is validated for use in ELISA, western blotting for the detection of Glial Derived Neurotrophic Factor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glial Derived Neurotrophic Factor
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified immunoglobulin. Liquid in PBS containing 0.02% sodium azide.
Gene Symbols GDNF; HFB1-GDNF; ATF1; ATF2; hGDNF; ATF
Host Species Chicken
Immunogen Synthetic peptides from human GDNF.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_000514.2; NM_199231.1; NM_199234.1
Target Species Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.