Learn More
Glial Derived Neurotrophic Factor Sheep anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Glial Derived Neurotrophic Factor |
| Applications | Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Lyophilized. Reconstitute with 50μL of sterile distilled water. Centrifuge to remove any residue. Contains no preservative. Glycerol (1:1) can be added for additional stability. |
| Gene Symbols | GDNF; HFB1-GDNF; ATF1; ATF2; hGDNF; ATF |
| Host Species | Sheep |
| Immunogen | Recombinant human GDNF. |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.