Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glial Fibrillary Acidic Protein Mouse anti-Chicken, Human, Pig, Rat, AlexaFluor™488, Clone: GA5, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB3402X
Description
Detect Glial Fibrillary Acidic Protein using this Anti-Glial Fibrillary Acidic Protein Antibody, clone GA5, Alexa Fluor 488 validated for use in immunocytochemistry, immunohistochemistry.
Specifications
| Glial Fibrillary Acidic Protein | |
| Monoclonal | |
| Alexa Fluor 488 | |
| NM_002055.2 | |
| Mouse | |
| 100 μg | |
| Chicken, Human, Pig, Rat | |
| Purified |
| Immunocytochemistry, Immunohistochemistry | |
| GA5 | |
| Purified immunoglobulin conjugated to Alexa Fluor™ 488. Liquid in Phosphate buffer with 15mg/mL BSA as a stabilizer and 0.1% sodium azide. | |
| GFAP, FLJ45472 | |
| Purified glial filament (1). | |
| Primary | |
| 2-8°C in the dark for up to 6 months. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction