Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glial Fibrillary Acidic Protein Mouse anti-Human, Rat, Mouse, Pig, Chicken, Bovine, Rabbit, Clone: GA5, MilliporeSigma™

Catalog No. MAB3402MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
40 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3402MI 40 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3402MI Supplier MilliporeSigma Supplier No. MAB3402
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Glial Fibrillary Acidic Protein Antibody, clone GA5 is an antibody against Glial Fibrillary Acidic Protein for use in immunocytochemistry, immunohistochemistry, immunohistochemistry (paraffin) and western blotting with more than 55 product citations.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glial Fibrillary Acidic Protein
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone GA5
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1 in buffer containing 0.02Mphosphate buffer, 0.25MNaCl with 0.1% sodium azide, pH 7.6.
Gene Accession No. NM_002055.2
Gene Symbols GFAP, FLJ45472
Host Species Mouse
Immunogen Purified glial filament (Debus, E., 1983).
Purification Method Protein A purfied
Quantity 40 μg
Primary or Secondary Primary
Target Species Bovine, Chicken, Human, Mouse, Pig, Rabbit, Rat
Content And Storage 2-8°C in undiluted aliquots for up to 6 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.