Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Glial Fibrillary Acidic Protein (GFAP) Clone: 2.2B10,

Catalog No. 345860100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
34-586-0100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-586-0100UG Supplier MilliporeSigma™ Supplier No. 345860100UG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Specifically detects Glial Fibrillary Acidic Protein (GFAP) Clone: 2.2B10 in Bovine, Human, Rodent samples, and it is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Immuno Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Glial Fibrillary Acidic Protein (GFAP)
Applications ELISA, Immunoblot, Immunoblot, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone 2.2B10
Conjugate Unconjugated
Formulation In PBS.
Host Species Rat
Immunogen bovine GFAP
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes GFAP in astrocytes induced by a variety of CNS injuries in human and rat brain tissues.
Target Species Bovine, Human, Rodent
Content And Storage −20°C
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.