Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Glial Fibrillary Acidic Protein (GFAP) Clone: G-A-5,

Catalog No. IF03L100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
IF03L100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. IF03L100UG Supplier MilliporeSigma™ Supplier No. IF03L100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Glial Fibrillary Acidic Protein (GFAP) Clone: G-A-5 in Chicken, Human, Porcine, Rat samples, and it is validated for Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Immuno Blotting

Intermediate (10 nm) filaments are present in, and provide information concerning the origin of, most vertebrate cells. At present, five major cell types can be distinguished by their intermediate filament components. These include epithelial cells (cytokeratins), neurons (neurofilaments), glial cells (glial fibrillary acidic protein or GFA or GFAP), skeletal, visceral, and certain vascular smooth muscle cells (desmin) and various nonepithelial cells, including cells of mesenchymal origin (vimentin). GFAP and vimentin are coexpressed in at least some astrocytes. Anti-GFAP (Ab-1) identifies the GFAP subclass of intermediate filaments in human tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glial Fibrillary Acidic Protein (GFAP)
Applications Immunoblot, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Clone G-A-5
Conjugate Unconjugated
Formulation Lyophilized from 20mM NH4HCO3, 100μg BSA.
Host Species Mouse
Immunogen GFAP
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes GFAP in astrocytes, astrocytomas, glial cells, gliomas, and other glial cell-derived tumors.
Target Species Chicken, Human, Pig, Rat
Content And Storage 2°C to 8°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.