Learn More
Glial Fibrillary Acidic Protein (GFAP) Rabbit anti-Bovine, Canine, Human, Rat, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Glial Fibrillary Acidic Protein (GFAP) |
| Applications | Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Rabbit serum. Liquid. Contains no preservative. |
| Gene Symbols | FLJ45472; GFAP |
| Host Species | Rabbit |
| Immunogen | Purified bovine GFAP |
| Purification Method | Unpurified |
| Quantity | 50 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.