Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glial Fibrillary Acidic Protein Mouse anti-Human, Phycoerythrin, Clone: GA5, MilliporeSigma™

Catalog No. FCMAB257PMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FCMAB257PMI 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FCMAB257PMI Supplier MilliporeSigma Supplier No. FCMAB257P
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Milli-Mark™Anti-Glial Fibrillary Acidic Protein-PE Antibody, clone GA5 detects level of Glial Fibrillary Acidic Protein and has been published and validated for use in flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glial Fibrillary Acidic Protein
Applications Flow Cytometry
Classification Monoclonal
Clone GA5
Conjugate PE
Formulation Purified mouse monoclonal IgG1 conjugated to PE in PBS with 0.1% sodium azide and 15mg/mL BSA.
Gene Accession No. NP_002046
Gene Symbols GFAP, FLJ45472
Host Species Mouse
Immunogen Purified glial filament (Debus, E., 1983).
Purification Method Protein A Purified
Quantity 100 Tests
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for up to 6 months. Protect from light.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.