Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glial Fibrillary Acidic Protein Chicken anti-Human, Mouse, Rat, Bovine, Pig, Polyclonal, MilliporeSigma™
GSA_VA

Catalog No. AB5541MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5541MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5541MI Supplier MilliporeSigma Supplier No. AB5541
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

This Anti-Glial Fibrillary Acidic Protein Antibody is validated for use in immunocytochemistry, immunohistochemistry, immunohistochemistry (paraffin), western blotting for the detection of GFAP.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Glial Fibrillary Acidic Protein
Applications Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified chicken polyclonal in buffer containing PBS containing 5mM sodium azide.
Gene Symbols GFAP; FLJ45472
Host Species Chicken
Immunogen Purified bovine GFAP.
Purification Method Affinity Purified
Quantity 50 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_002055.2
Target Species Bovine, Human, Mouse, Pig, Rat
Content And Storage -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.