Learn More
GLI pathogenesis-related 1, Mouse, Polyclonal Antibody, Abnova™
Shop All Abnova Corporation ProductsDescription
This gene encodes a protein with similarity to both the pathogenesis-related protein (PR) superfamily and the cysteine-rich secretory protein (CRISP) family. Increased expression of this gene is associated with myelomocytic differentiation in macrophage and decreased expression of this gene through gene methylation is associated with prostate cancer. The protein has proapoptotic activities in prostate and bladder cancer cells. This gene is a member of a cluster on chromosome 12 containing two other similar genes. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized. [provided by RefSeq
Specifications
Specifications
| Antigen | GLI pathogenesis-related 1 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Mouse polyclonal antibody raised against a partial recombinant GLIPR1. |
| Formulation | 50% glycerol |
| Gene | GLIPR1 |
| Gene Accession No. | NM_006851 |
| Gene Alias | CRISP7/GLIPR/RTVP1 |
| Gene Symbols | GLIPR1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.