Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLP-1 (7-36) Rabbit anti-Mammals, Polyclonal, MilliporeSigma™

Catalog No. AB3244MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB3244MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB3244MI Supplier MilliporeSigma Supplier No. AB3244
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-GLP-1 (7-36) Antibody, C-terminus amide-reactive detects level of GLP-1 (7-36) and has been published and validated for use in ELISA, RIA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLP-1 (7-36)
Applications ELISA, Radioimmune Assays (RIA)
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit serum, diluted. Liquid in PBS containing 0.05% sodium azide.
Gene Symbols GCG; GLP1; glucagon; GRPP; GLP2
Host Species Rabbit
Immunogen Whole synthetic peptide.
Quantity 50 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_002054.2
Target Species Mammalia
Content And Storage −20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.