Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLP1R Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. AB9433IMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB9433IMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB9433IMI Supplier MilliporeSigma Supplier No. AB9433I
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-GLP1R Antibody is validated for use in Immunohistochemistry (Paraffin) for the detection of GLP1R.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLP1R
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit Polyclonal serum in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols GLP1R
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to the extracellular domain of Human Glucagon-Like Peptide 1 Receptor.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_002053
Target Species Human, Mouse
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.