Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

glutaminase, Mouse, Clone: 5C4, Abnova™

Catalog No. 89014117 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-014-117 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-014-117 Supplier Abnova Corporation Supplier No. H00002744M01
Only null left
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant GLS.

Sahai (1983) [PubMed 6825316] demonstrated phosphate-activated glutaminase (EC 3.5.1.2) in human platelets. It is the major enzyme yielding glutamate from glutamine. Significance of the enzyme derives from its possible implication in behavior disturbances in which glutamate acts as a neurotransmitter (Prusiner, 1981). High heritability of platelet glutaminase was indicated by studies of Sahai and Vogel (1983) [PubMed 6682827] who found an intraclass correlation coefficient of 0.96 for monozygotic twins and 0.53 for dizygotic twins.[supplied by OMIM

Sequence: EQRDYDSRTALHVAAAEGHVEVVKFLLEACKVNPFPKDRWNNTPMDEALHFGHHDVFKILQEYQVQYTPQGDSDNGKENQTVHKNLDGLL

Specifications

Antigen glutaminase
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 5C4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant GLS.
Formulation PBS with no preservative; pH 7.4
Gene GLS
Gene Accession No. NM_014905
Gene Alias AAD20/DKFZp686O15119/FLJ10358/GLS1/KIAA0838
Gene Symbols GLS
Host Species Mouse
Immunogen GLS (NP_055720, 580 a.a. ∼ 669 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2744
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.