Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
glioma tumor suppressor candidate region gene 2, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant GLTSCR2.
Supplier: Abnova Corporation H00029997A01
Description
Sequence: KPRRLGRLKYQAPDIDVQLSSELTDSLRTLKPEGNILRDRFKSFQRRNMIEPRERAKFKRKYKVKLVEKRAFREIQLSpecifications
glioma tumor suppressor candidate region gene 2 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant GLTSCR2. | |
GLTSCR2 | |
PICT-1/PICT1 | |
Mouse | |
50 μL | |
Yes | |
29997 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_015710 | |
GLTSCR2 | |
GLTSCR2 (NP_056525, 402 a.a. ∼ 478 a.a) partial recombinant protein with GST tag. | |
RUO | |
Primary | |
Human | |
Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction