Learn More
Glucagon Receptor Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA527183
Description
PA5-27183 targets Glucagon Receptor in IHC (P) and WB applications and shows reactivity with Human samples. The PA5-27183 immunogen is synthetic peptide corresponding to a region within amino acids 36 and 130 of Human Glucagon Receptor. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
The physiologic effects of glucagon (GCG; MIM 138030) are mediated through the glucagon receptor, a member of the superfamily of receptors characterized by a 7-transmembrane domain structure and by their coupling via GTP-binding proteins (G proteins) to adenyl cyclase.
Specifications
Glucagon Receptor | |
Polyclonal | |
Unconjugated | |
P47871 | |
GCGR | |
Synthetic peptide corresponding to a region within amino acids 36 and 130 of Human Glucagon Receptor | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
GCGR | |
GGR, GL-R | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2642 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.