Learn More
anti-Glucose Regulated Protein 78 (GRP78) (BiP), Polyclonal, Invitrogen™
Applications: IF, IP, WB
Supplier: Thermo Scientific PA1014
Description
The 78 kDa glucose regulated protein/BiP (GRP78) belongs to the family of ∽70 kDa heat shock proteins (HSP70). GRP78 is a resident protein of the endoplasmic reticulum (ER) and associates transiently with a variety of newly synthesized secretory and membrane proteins. GRP78 may also interact with mutant and misfolded proteins, marking them for degredation. The highly conserved sequence Lys-Asp-Glu-Leu (KDEL) is present at the C-terminus of GRP78 and other resident ER proteins including glucose regulated protein 94 (GRP94) and protein disulfide isomerase (PDI). The KDEL signal is recognized by the KDEL receptor and this interaction ensures recycling of escaped ER proteins back to the ER.
Host Species: Rabbit
Cross Reactivity: Ms, Rt
Western Blotting

Specifications
Glucose Regulated Protein 78 (GRP78) (BiP) | |
Unconjugated | |
100 μg | |
Mouse, Rat |
Polyclonal | |
Rabbit | |
Primary | |
Purified |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.