Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glucose Regulating Protein 94 (GRP94) Rabbit anti-Human, Monkey, Rat, Polyclonal, MilliporeSigma™

Catalog No. 071408MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-140-8MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-140-8MI Supplier MilliporeSigma Supplier No. 071408
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Glucose Regulating Protein 94 (GRP94) Antibody detects level of Glucose Regulating Protein 94 (GRP94) and has been published and validated for use in western blotting and immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glucose Regulating Protein 94 (GRP94)
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit polyclonal IgG serum in buffer containing 0.02% sodium azide.
Gene Symbols HSP90B1; GP96; GRP94; TRA1; ECGP
Host Species Rabbit
Immunogen Peptide corresponding to an internal sequence from human GRP94.
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_003299.1
Target Species Human, Monkey, Rat
Content And Storage -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.