Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Glucose Transporter-3, Polyclonal

Catalog No. 40006250UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
40-006-250UG 50 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-006-250UG Supplier MilliporeSigma™ Supplier No. 40006250UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects Glucose Transporter-3 Clone: in Human samples, and it is validated for ELISA, Immunofluorescence, Immunohistochemistry (Frozen), Immunoprecipitation, Immuno Blotting

Glucose transporter-3 (Glut-3) is a transmembrane protein that facilitates transport of glucose in the cell in an energy-independent fashion. Glut-3 is believed to be the major neuronal glucose transporter. The tissue distribution of human Glut-3 protein appears to be primarily restricted to the brain and testis/spermatozoa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glucose Transporter-3
Applications ELISA, Immunoblot, Immunoblot, Immunofluorescence, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Formulation In PBS, 0.1% BSA, pH 7.5.
Host Species Rabbit
Immunogen a synthetic peptide corresponding to amino acids near the C-terminus of human GLUT-3, conjugated to KLH
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽45kDa GLUT-3 protein.
Target Species Human
Content And Storage −20°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.