Learn More
Glucose Transporter GLUT-1 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Glucose Transporter GLUT-1 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | The polyclonal is presented in phosphate buffered saline containing 10mM sodium azide. We recommend that each laboratory determine an optimum working titre for use in its particular application. |
| Gene Symbols | SLC2A1; GLUT-1; MGC141896; GLUT1; MGC141895; GLUT |
| Host Species | Rabbit |
| Immunogen | 12 amino acid peptide of the C-terminus of hGLUT-1 {LFHPLGADSQV} conjugated to KLH |
| Purification Method | Affinity Purified |
| Quantity | 100 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.