Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glucose Transporter GLUT-4 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. CBL243MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CBL243MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CBL243MI Supplier MilliporeSigma Supplier No. CBL243
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-Glucose Transporter GLUT-4 Antibody is validated for use in immunoprecipitation, western blotting for the detection of Glucose Transporter GLUT-4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glucose Transporter GLUT-4
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified immunoglobulin in PBS with 0.01% sodium azide.
Gene Symbols SLC2A4; GLUT-4; GLUT4
Host Species Rabbit
Immunogen Partially purified GLUT-4
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_001042.2
Target Species Human, Mouse, Rat
Content And Storage 4°C for 1 month, −20°C in undiluted aliquotes for long term
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.