Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GluR1 Rabbit anti-Human, Mouse, Rat, Chimpanzee, Clone: C3T, MilliporeSigma™

Catalog No. 04855MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-855-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-855-MI Supplier MilliporeSigma Supplier No. 04855
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Anti-GluR1 Antibody, clone C3T detects level of GluR1 and has been published and validated for use in immunohistochemistry, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GluR1
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone C3T
Conjugate Unconjugated
Formulation Cultured supernantant in 0.05% sodium azide
Gene Accession No. NM_000827.2
Host Species Rabbit
Immunogen KLH-conjugated, synthetic peptide corresponding to amino acids 858-868 (..TLPRNSGAGAS..) of human GluR1 (amino acids 840-850 of mature human GluR1)
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) GRIA1, GluR-A, GluR-1, GLUR1, GluR-K1, GLURA, HBGR1, GLUH1, MGC133252
Target Species Chimpanzee, Human, Mouse, Rat
Content And Storage −20°C for 24 months. Aliquot to avoid freeze/thaw cycles.
Form Supernatant
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.