Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GluR1-NT (NT) Mouse anti-Rat, Clone: RH95, MilliporeSigma™

Catalog No. MAB2263MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2263MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB2263MI Supplier MilliporeSigma Supplier No. MAB2263
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-GluR1-NT (N-terminus) Antibody, clone RH95 is an antibody against GluR1-NT (N-terminus) for use in western blot, immunoprecipitation, immunocytochemistry, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GluR1-NT (NT)
Applications ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone RH95
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
Gene Accession No. NP_113796.1
Gene Symbols GLUH1, GLUR1, GLURA, GluR-1, GluR-A, GluR-K1, HBGR1, MGC133252, GRIA1
Host Species Mouse
Immunogen Thyroglobulin-conjugated linear peptide of rat GluR1 designed around the N-terminus.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.