Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GluR1 Rabbit anti-Mouse, Rat, Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABN241
Description
Anti-GluR1 Antibody is a highly specific rabbit polyclonal antibody, that targets GluR1 and has been tested in western blotting and immunohistochemistry (Paraffin).
Specifications
GluR1 | |
Polyclonal | |
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
NP_113796 | |
KLH-conjugated linear peptide corresponding to the extracellular domain of rat GluR1. | |
100 μg | |
Gria1, Glur1 | |
2-8°C for 12 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction