Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GluR1 Rabbit anti-Mouse, Rat, MilliporeSigma™

Catalog No. 05855RMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-855-RMI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-855-RMI Supplier MilliporeSigma Supplier No. 05855R
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Anti-GluR1 Antibody, Recombinant is an antibody against GluR1 for use in western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GluR1
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Purified Antibody in PBS, 0.05% NaN3
Gene Accession No. NP_000818.2
Host Species Rabbit
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) GLUH1, GLUR1, GLURA, GluR-1
Target Species Mouse, Rat
Content And Storage 2-8°C in undiluted aliquots for up to 12 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.