Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GluR2 Mouse anti-Rat, Mouse, Human, Clone: L21/32, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABN71
Description
Detect GluR2 using this Anti-GluR2 Antibody, clone L21/32 validated for use in western blot, immunohistochemistry.
Specifications
| GluR2 | |
| Monoclonal | |
| Unconjugated | |
| NP_000817 | |
| Mouse | |
| Protein G | |
| Primary | |
| 2-8°C for one year | |
| IgG1 κ |
| Immunohistochemistry, Western Blot | |
| L21/32 | |
| Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
| HBGR2, GLUR2, GLURB, GluR-K2, GluR-B, GluR-2, gluR-B, GRIA2 | |
| Recombinant protein corresponding to human GluR2. | |
| 100 μg | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction