Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GluR2 Mouse anti-Human, FITC, Clone: 6C4, MilliporeSigma™

Catalog No. FCMAB290FMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FCMAB290FMI 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FCMAB290FMI Supplier MilliporeSigma Supplier No. FCMAB290F

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

This Milli-Mark™ Anti-GluR2-FITC Antibody, clone 6C4 is validated for use in FC for the detection of GluR2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GluR2
Applications Flow Cytometry
Classification Monoclonal
Clone 6C4
Conjugate FITC
Formulation Purified mouse monoclonal IgG2a conjugated to FITC in PBS with 0.1% sodium azide and 15mg/mL BSA.
Gene Accession No. NP_001077280
Gene Symbols Gria2, GLUR2, GLURB, GluR-2, GluR-B, GluR-K2
Host Species Mouse
Immunogen Recombinant fusion protein TrpE-GluR2 (putative N-terminal portion, amino acids 175-430).
Purification Method Protein A Purified
Quantity 100 Tests
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for up to 6 months. Protect from light.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.