Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GLUR2 pS880 Mouse anti-Rat, Clone: 022.19.5, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABN103
Description
This Anti-GLUR2 pS880 (Clone 022.19.5) Antibody is validated for use in western blotting for the detection of GLUR2 pS880 (Clone 022.19.5).
Specifications
GLUR2 pS880 | |
Monoclonal | |
Unconjugated | |
NP_001077280 | |
Mouse | |
Protein G | |
Primary | |
2-8°C for one year | |
IgG2a κ |
Western Blot | |
022.19.5 | |
Purified mouse monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
Gria2, Glur2 | |
KLH-conjugated linear peptide corresponding to rat GluR2 phosphorylated at Ser880. | |
100 μg | |
Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction