Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GluR6/7 Rabbit anti-Human, Mouse, Rat, Clone: NL904, MilliporeSigma™
Rabbit Monoclonal Antibody
Supplier: MilliporeSigma 04921
Description
Anti-GluR6/7 Antibody, clone NL9 is an antibody against GluR6/7 for use in immunoprecipitation and western blotting.
Specifications
GluR6/7 | |
Monoclonal | |
Unconjugated | |
Rabbit | |
None | |
Primary | |
Human, Mouse, Rat | |
Supernatant |
Immunoprecipitation, Western Blot | |
NL904 | |
NM_021956 | |
KLH-conjugated, synthetic peptide corresponding to amino acids 894-908 (C-terminus) of rat GluR6. The immunizing sequence is identical in human. | |
100 μL | |
GRIK2, GLR6, EAA4, GluR-6, GLUR6, MGC74427, GluR6 | |
−20°C for 24 months. Aliquot to avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction