Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLUT-1 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
GSA_VA

Catalog No. 071401MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-140-1MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-140-1MI Supplier MilliporeSigma Supplier No. 071401
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-GLUT-1 Antibody, C-terminus is an antibody against C terminus of GLUT-1 that is validated for the detection of GLUT-1 in Western Blotting (WB), Immunohistochemistry - Paraffin (immunohistochemistry-P), Immunocytochemistry (immunocytochemistry) and ELISA.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Antigen GLUT-1
Applications ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal IgG in buffer containing 0.02M Phosphate Buffer, 0.25M NaCl, with 0.1% sodium azide.
Gene Symbols GLUT; GLUT-1; GLUT1; MGC141895; MGC141896
Host Species Rabbit
Immunogen Synthetic peptide corresponding amino acids with the C-terminus of human GLUT-1 coupled to KLH (C-ELFHPLGADSQV).
Purification Method Affinity Purified
Quantity 50 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_006516
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for 12 months
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.