Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLUT-4 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. 071404MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-140-4MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-140-4MI Supplier MilliporeSigma Supplier No. 071404
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect GLUT-4 using this Anti-GLUT-4 Antibody, C-terminus validated for use in western blot, immunohistochemistry (paraffin), ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLUT-4
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing PBS with 0.05% sodium azide.
Gene Symbols SLC2A4; GLUT-4; GLUT4
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to the C-terminus of human GLUT-4.
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_001033
Target Species Human, Mouse
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.