Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLUT-5 Rabbit anti-Rat, Mouse, Guinea Pig, Polyclonal, MilliporeSigma™

Catalog No. 071406MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-140-6MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-140-6MI Supplier MilliporeSigma Supplier No. 071406
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-GLUT-5 Antibody, a.a. 490-502 detects level of GLUT-5 and has been published and validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GLUT-5
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Liquid containing 0.05% sodium azide.
Gene Symbols GLUT-5; GLUT5; OTTHUMP00000001704
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the C-terminus of rat GLUT-5 (aa 490-502) coupled to KLH. The immunogen sequence used is only 53% conserved in mouse.
Purification Method Unpurified rabbit serum
Quantity 50 μL
Primary or Secondary Primary
Gene ID (Entrez) P22732
Target Species Guinea Pig, Mouse, Rat
Content And Storage -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.