Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutamate Receptor 1 Rabbit anti-Rat, Polyclonal, MilliporeSigma™

Catalog No. AB584925UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB584925UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB584925UL Supplier MilliporeSigma Supplier No. AB584925UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-Glutamate Receptor 1 Antibody, phosphoSer 845 is validated for use in western blotting for the detection of Glutamate Receptor 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glutamate Receptor 1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified immunoglobulin. Liquid in 10mM HEPES, pH 7.5, with 150mM NaCl, 100μg/mL BSA and 50% glycerol.
Gene Symbols GRIA1; GluR-K1; GLURA; HBGR1; GluR-A; GluR-1; GLUH1; GLUR1; MGC133252
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids surrounding the phosphoSer845 of rat GluR1.
Purification Method Affinity Purified
Quantity 25 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_000827.2
Target Species Rat
Content And Storage −20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.