Learn More
Glutamate receptor 1 Rabbit anti-Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB1504
Description
This Anti-Glutamate receptor 1 Antibody is validated for use in western blot, immunohistochemistry (paraffin), immunoprecipitation for the detection of Glutamate receptor 1.
Specifications
Glutamate Receptor 1 | |
Polyclonal | |
Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
Equine, Human, Mouse, Rat |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
GRIA1; GluR-K1; GluR-A; GLUH1; GLUR1; GLURA; HBGR1; GluR-1 | |
KLH-conjugated linear peptide corresponding to human Glutamate receptor 1 at the cytoplasmic domain. | |
200 μL | |
NP_000818.2 | |
2-8°C for 12 months |