Learn More
Glutamate Receptor 2 & 3 Rabbit anti-Human, Rat, Mouse, Monkey, Guinea Pig, Hamster, Gerbil, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Glutamate Receptor 2 & 3 |
| Applications | Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Purified rabbit polyclonal serum lyophilized from buffer containing 0.01M phosphate, 0.09M NaCl, pH 7.2. 1% BSA and 0.1% sodium azide. |
| Gene Symbols | GRIA2; GluR-B; gluR-B; GluR-2; GLUR2; GLURB; HBGR2; GluR-K2 |
| Host Species | Rabbit |
| Immunogen | Carboxy terminus peptide of rat GluR2 conjugated to BSA with glutaraldehyde (EGYNVYGIESVKI). |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.