Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutamate Receptor 3 Mouse anti-Human, Mouse, Rat, Monkey, Clone: 3B3, MilliporeSigma™

Catalog No. MAB5416MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5416MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB5416MI Supplier MilliporeSigma Supplier No. MAB5416
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Glutamate Receptor 3 Antibody, clone 3B3 is an antibody against Glutamate Receptor 3 for use in western blot, immunocytochemistry, immunohistochemistry, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glutamate Receptor 3
Applications Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 3B3
Conjugate Unconjugated
Formulation Liquid in PBS. Contains no preservative.
Gene Accession No. NM_000840.2
Gene Symbols GLUR3, GluR3, GLURC, GRIA3
Host Species Mouse
Immunogen Fusion protein, N-terminus of GluR3
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Monkey, Mouse, Rat
Content And Storage Stable for 1 year at from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.